Skip to Content
Merck
All Photos(1)

Key Documents

738875

Sigma-Aldrich

Allyl oxalyl chloride

97%

Synonym(s):

2-Propen-1-yloxy chloroacetate, Allyl 2-chloro-2-oxoacetate, Chlorooxoacetic acid 2-propenyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$45.60
5 G
$145.00

$45.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$45.60
5 G
$145.00

About This Item

Empirical Formula (Hill Notation):
C5H5ClO3
CAS Number:
Molecular Weight:
148.54
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$45.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.443

density

1.213 g/mL at 25 °C

functional group

acyl chloride
ester

SMILES string

ClC(=O)C(=O)OCC=C

InChI

1S/C5H5ClO3/c1-2-3-9-5(8)4(6)7/h2H,1,3H2

InChI key

HNOLIWBAJVIBOU-UHFFFAOYSA-N

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

133.0 °F - closed cup

Flash Point(C)

56.1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service