Skip to Content
Merck
All Photos(3)

Key Documents

185248

Sigma-Aldrich

Allyl acetate

99%

Synonym(s):

3-Acetoxy-1-propene

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$92.80
500 ML
$256.00

$92.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$92.80
500 ML
$256.00

About This Item

Linear Formula:
CH3COOCH2CH=CH2
CAS Number:
Molecular Weight:
100.12
Beilstein:
1742050
EC Number:
MDL number:
UNSPSC Code:
12162002
eCl@ss:
39022118
PubChem Substance ID:
NACRES:
NA.23

$92.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

autoignition temp.

705 °F

refractive index

n20/D 1.404 (lit.)

bp

103-104 °C (lit.)

density

0.928 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)OCC=C

InChI

1S/C5H8O2/c1-3-4-7-5(2)6/h3H,1,4H2,2H3

InChI key

FWZUNOYOVVKUNF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service