Skip to Content
Merck
All Photos(3)

Key Documents

929654

Sigma-Aldrich

ElectroGreen®

greener alternative

Isopropyl Alcohol substitute for electronics, bio-based

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$78.80

$78.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
$78.80

About This Item

UNSPSC Code:
12190000
NACRES:
NA.05

$78.80


Check Cart for Availability

Request a Bulk Order

Quality Level

description

Relative Evaporation rate: 1.24


Hansen solubility parameters: SPd = 7.7; SPp = 4.2; SPh = 8.8.

for electronic purposes

Assay

≥ 99.0% (GC)

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

impurities

≤0.09 wt. % Acidity (as lactic acid)
≤0.5 wt. % Water (Karl Fischer)
≤1 ppm As, Cr, Cd, Cu, Hg, Mn, Ni, Pb, Zn, trace (ICP, each)

evapn. residue

≤0.05%

color

clear

viscosity

9.8  cP(20 °C)

density

0.8351 at 25 °C (specific gravity)

greener alternative category

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

This ElectroGreen® solvent is made of bio-based (corn/beets-derived)safer alternatives to replace synthetic propanol/IPA fossil-based solvent with specifications tailored for electronics and energy applications. All our ElectroGreen® solvents line are derived from a renewable resource (verified through C14 ASTM D6866-16 testing). We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Click here for more details.

This solvent blend consists of ethanol 70-90%, Ethyl lactate 20-30%, ethyl acetate 1-5%.

Application

Our Isopropyl Alcohol substitute is a blend of bio-based solvents that provide excellent cleaning properties on electronics: dries quickly, and provides a clean, non-residue surface. Ideal for dilution in a hot water spray at time of use. Suitable replacement for Isopropyl Alcohol (IPA).

Storage and Stability

To ensure low water content, handle under inert conditions. Product should be refrigerated upon receipt.

Legal Information

ElectroGreen is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

52.0 °F

Flash Point(C)

11.1 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Carbon-based Sustainable Organic Electronics (SOE) limit the use of critical elements and biodegrade at their end-of-life. This review offers insight on how structural and energy disorder in these materials influence device performance and includes evaluations of various transport models and their limitations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service