Skip to Content
Merck
All Photos(3)

Key Documents

929689

Sigma-Aldrich

ElectroGreen®

greener alternative

Toluene substitute for electronics, bio-sourced

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$84.70

$84.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
$84.70

About This Item

UNSPSC Code:
12190000
NACRES:
NA.06

$84.70


Check Cart for Availability

Request a Bulk Order

description

Relative Evaporation rate: 2.57
Hansen Solubility Parameters: SPd = 7.7; SPp = 2.8; SPh = 6.

for electronic purposes

Quality Level

Assay

≥99%

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

impurities

≤0.05 wt. % acidity (as lactic acid)
≤0.2 wt. % water (Karl Fischer)
≤1 ppm As, Cr, Cd, Cu, Hg, Mn, Ni, Pb, Zn, trace (ICP)

evapn. residue

≤0.05%

color

clear

viscosity

8.1  cP(20 °C)

density

0.965 at 25 °C (specific gravity)

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

This ElectroGreen® solvent is made of bio-based (corn/beets-derived) safer alternatives to replace synthetic toluene fossil-based solvent with specifications tailored for electronics and energy applications. All our ElectroGreen® solvents line are derived from a renewable resource (verified through C14 ASTM D6866-16 testing). We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Click here for more details.

This solvent blend consists of ethanol 20-30%, Ethyl lactate 20-30%, ethyl acetate 10-20%.

Application

Toluene is used in electronics industry as cleaning solvent and in ink formulations. Our toluene substitute is a bio-based blend replacement for toluene. With our toluene substitute, ink formulators are reducing the use of hazardous petroleum solvents.

Storage and Stability

To ensure low water content, handle under inert conditions. Product should be refrigerated upon receipt.

Legal Information

ElectroGreen is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

44.6 °F

Flash Point(C)

7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Carbon-based Sustainable Organic Electronics (SOE) limit the use of critical elements and biodegrade at their end-of-life. This review offers insight on how structural and energy disorder in these materials influence device performance and includes evaluations of various transport models and their limitations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service