Skip to Content
Merck
All Photos(1)

Key Documents

922390

Sigma-Aldrich

Poly (2-(N-3-Sulfopropyl-N,N-dimethyl ammonium)ethyl methacrylate)

Mn 20,000

Synonym(s):

PDMAPS, PSBMA

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
$305.00

$305.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 G
$305.00

About This Item

Linear Formula:
(C11H21NO5S)n
UNSPSC Code:
12162002
NACRES:
NA.23

$305.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

mol wt

Mn 20,000

Application

Poly(sulfobetaine methacrylate) (PSBMA) is a well-known zwitterionic polymer, it is non-toxic and biocompatible. On each of its repeating unit, it carries both a positive charge and a negative charge, leading to its strong hydration capacity. Surfaces coated with PSBMA showed superhydrophilic property and strong resistance to protein adsorption/bacterial adhesion, making this polymer a promising material in anti-fouling applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reza Lalani et al.
Biomacromolecules, 13(6), 1853-1863 (2012-05-02)
Zwitterionic poly(sulfobetaine methacrylate) (PSBMA) has been well studied for its superhydrophilic and ultralow biofouling properties, making it a promising material for superabsorbent and nonadherent wound dressings. Electrospinning provides multiple desirable features for wound dressings, including high absorptivity due to high
Se Yeong Lee et al.
Biomaterials research, 22, 3-3 (2018-02-17)
Zwitterionic molecules have been widely studied as coating materials for preparing anti-fouling surfaces because they possess strong hydration properties that can resist non-specific protein adsorption. Numerous studies on surface modification using zwitterionic molecules have been investigated, such as electrochemically mediated

Questions

  1. For product 922749 how it is end capped?

    1 answer
    1. Product 922749  Poly(2-methacryloyloxyethyl phosphorylcholine) is end capped with hydroxy (OH) group similar to 925632.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service