Skip to Content
Merck
All Photos(3)

Key Documents

908703

Sigma-Aldrich

[Ir(p-F(Me)ppy)2-(4,4′-dtbbpy)]PF6

≥95%

Synonym(s):

[(4,4′-di-tert-butyl-2,2′-bipyridine)-bis-(5-methyl-2-(5-fluoro-phenyl)-pyridine)-iridium(III)] hexafluorophosphate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$254.00
500 MG
$826.00

$254.00


Estimated to ship on29 March 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$254.00
500 MG
$826.00

About This Item

Empirical Formula (Hill Notation):
C42H42F8IrN4P
CAS Number:
Molecular Weight:
977.99
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

$254.00


Estimated to ship on29 March 2025


Request a Bulk Order

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

photocatalyst activation

450 nm

Application

[Ir(p-F(Me)ppy)2-(4,4′-dtbbpy)]PF6 is a cyclometalated iridium(III) complex that can be used in visible-light mediated photocatalytic organic transformations.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yufan Liang et al.
Nature, 559(7712), 83-88 (2018-06-22)
Over the past three decades, considerable progress has been made in the development of methods to construct sp2 carbon-nitrogen (C-N) bonds using palladium, copper or nickel catalysis1,2. However, the incorporation of alkyl substrates to form sp3 C-N bonds remains one
Andrew G Capacci et al.
Nature chemistry, 9(11), 1073-1077 (2017-10-25)
Although the α-alkylation of ketones has already been established, the analogous reaction using aldehyde substrates has proven surprisingly elusive. Despite the structural similarities between the two classes of compounds, the sensitivity and unique reactivity of the aldehyde functionality has typically
Yong Yao Loh et al.
Science (New York, N.Y.), 358(6367), 1182-1187 (2017-11-11)
Deuterium- and tritium-labeled pharmaceutical compounds are pivotal diagnostic tools in drug discovery research, providing vital information about the biological fate of drugs and drug metabolites. Herein we demonstrate that a photoredox-mediated hydrogen atom transfer protocol can efficiently and selectively install

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service