Skip to Content
Merck
All Photos(1)

Key Documents

900570

Sigma-Aldrich

Ir[dF(t-Bu)-ppy]3

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$144.00

$144.00


Estimated to ship on31 March 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$144.00

About This Item

Empirical Formula (Hill Notation):
C45H42F6IrN3
CAS Number:
Molecular Weight:
931.04
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

$144.00


Estimated to ship on31 March 2025


Request a Bulk Order

form

powder or crystals

Quality Level

reaction suitability

core: iridium
reagent type: catalyst
reaction type: Photocatalysis

mp

370.93 °C

photocatalyst activation

465 nm

SMILES string

FC1=CC([Ir]2(C3=C4C(F)=CC(F)=C3)([N]5=C4C=C(C(C)(C)C)C=C5)([N]6=C7C=C(C(C)(C)C)C=C6)[N]8=C(C9=C2C=C(F)C=C9F)C=C(C(C)(C)C)C=C8)=C7C(F)=C1

Application

Ir[dF(t-Bu)-ppy]3 is a photocatalyst that readily facilitates the decarboxylative arylation of α-amino acids using visible light.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service