Skip to Content
Merck
All Photos(3)

Key Documents

905917

Sigma-Aldrich

(Ir[Me(Me)ppy]2(dtbpy))PF6

Synonym(s):

4,4′-Bis(t-butyl-2,2′-bipyridine]bis[5-methyl-2-(4-methyl-2-pyridinyl-kN)phenyl-kC]iridium hexafluorophosphate, Ir(dmppy)2(dtbbpy)PF6

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$235.00
500 MG
$791.00

$235.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$235.00
500 MG
$791.00

About This Item

Empirical Formula (Hill Notation):
C44H48F6IrN4P
CAS Number:
Molecular Weight:
970.06
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

$235.00


Check Cart for Availability

Request a Bulk Order

form

powder

reaction suitability

core: iridium
reagent type: catalyst
reaction type: Photocatalysis

photocatalyst activation

465 nm

SMILES string

F[P-](F)(F)(F)(F)F.CC1=CC([Ir+]([N]2=C3C=C(C)C=C2)(C4=C5C=CC(C)=C4)([N]6=C7C=C(C(C)(C)C)C=C6)([N]8=C7C=C(C(C)(C)C)C=C8)[N]9=C5C=C(C)C=C9)=C3C=C1

Application

(Ir[Me(Me)ppy]2(dtbpy))PF6 or Ir(dmppy)2(dtbbpy)PF6 is an iridium photoredox catalyst that facilitates a variety of transformations using visible light, including the α- and β-alkylation of aldehydes.[1][2]

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct β -alkylation of aldehydes via photoredox organocatalysis.
Terrett JA, et al.
Journal of the American Chemical Society, 136(19), 6858-6861 (2014)
Direct, enantioselective α-alkylation of aldehydes using simple olefins.
Capacci AG, et al.
Nature Chemistry, 9(11), 1073-1073 (2017)
Zachary C Litman et al.
Nature, 560(7718), 355-359 (2018-08-17)
Living organisms rely on simultaneous reactions catalysed by mutually compatible and selective enzymes to synthesize complex natural products and other metabolites. To combine the advantages of these biological systems with the reactivity of artificial chemical catalysts, chemists have devised sequential
Jack A Terrett et al.
Journal of the American Chemical Society, 136(19), 6858-6861 (2014-04-24)
Direct β-alkylation of saturated aldehydes has been accomplished by synergistically combining photoredox catalysis and organocatalysis. Photon-induced enamine oxidation provides an activated β-enaminyl radical intermediate, which readily combines with a wide range of Michael acceptors to produce β-alkyl aldehydes in a
Andrew G Capacci et al.
Nature chemistry, 9(11), 1073-1077 (2017-10-25)
Although the α-alkylation of ketones has already been established, the analogous reaction using aldehyde substrates has proven surprisingly elusive. Despite the structural similarities between the two classes of compounds, the sensitivity and unique reactivity of the aldehyde functionality has typically

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service