903647
1-Heptyl-2-norbornene
≥95%
Synonym(s):
1-Heptylbicyclo[2.2.1]hept-2-ene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
≥95%
form
liquid
reaction suitability
reaction type: C-H Activation
reagent type: catalyst
Application
related product
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Nature chemistry, 10(8), 866-872 (2018-06-27)
Achieving site-selectivity in arene functionalization that is complementary to the site-selectivity from electrophilic aromatic substitution reactions has been a long-standing quest in organic synthesis. Palladium/norbornene cooperative catalysis potentially offers a unique approach to this problem, but its use has been
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service