Skip to Content
Merck
All Photos(1)

Key Documents

125482

Sigma-Aldrich

cis-Cyclooctene

contains 100-200 ppm Irganox 1076 FD as antioxidant, 95%

Synonym(s):

(Z)-Cyclooctene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14
CAS Number:
Molecular Weight:
110.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
95%

Quality Level

Assay

95%

form

liquid

contains

100-200 ppm Irganox 1076 FD as antioxidant

refractive index

n20/D 1.469 (lit.)

bp

145-146 °C (lit.)

mp

−16 °C (lit.)

solubility

H2O: insoluble

density

0.846 g/mL at 25 °C (lit.)

SMILES string

C1CCCC=CCC1

InChI

1S/C8H14/c1-2-4-6-8-7-5-3-1/h1-2H,3-8H2/b2-1-

InChI key

URYYVOIYTNXXBN-UPHRSURJSA-N

General description

cis-Cyclooctene undergoes addition reaction with carbon tetrachloride catalyzed by dichlorotris(triphenylphosphine) ruthenium(II) to yield 1-chloro-4-(trichloromethyl) cyclooctane (1,4-adduct). It undergoes epoxidation catalyzed by molybdenum oxide nanoparticles incorporated in a mesoporous silica shell coated on dense silica-coated magnetite nanoparticles.

Application

cis-Cyclooctene has been used to study the alkenes chemisorbed on silicon(100) by scanning tunneling microscopy.

Other Notes

Contains cyclooctane

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radical reactions in the coordination sphere. 4. Addition of carbon tetrachloride to cis-cyclooctene catalyzed by dichlorotris (triphenylphosphine) ruthenium (II).
Matsumoto H, et al.
The Journal of Organic Chemistry, 43(9), 1734-1736 (1978)
Voltage-dependent STM images of covalently bound molecules on Si (100).
Padowitz DF and Hamers RJ.
The Journal of Physical Chemistry B, 102(43), 8541-8545 (1998)
A magnetically recyclable nanocomposite catalyst for olefin epoxidation.
Mohammadreza Shokouhimehr et al.
Angewandte Chemie (International ed. in English), 46(37), 7039-7043 (2007-08-10)
Xiao-Song Xue et al.
Chemical reviews, 117(13), 8622-8648 (2017-03-11)
The most fundamental concepts in chemistry are structure, energetics, reactivity and their inter-relationships, which are indispensable for promoting chemistry into a rational science. In this regard, bond energy, the intrinsic determinant directly related to structure and reactivity, should be most
Anyang Peng et al.
Science advances, 6(5), eaax6637-eaax6637 (2020-02-18)
Traditionally, a catalyst functions by direct interaction with reactants. In a new noncontact catalytic system (NCCS), an intermediate produced by one catalytic reaction serves as an intermediary to enable an independent reaction to proceed. An example is the selective oxidation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service