Skip to Content
Merck
All Photos(2)

Key Documents

917745

Sigma-Aldrich

[Ni(dtbbpy)(H2O)4]Cl2

Synonym(s):

Nickel(2+), tetraaqua[4,4′-bis(1,1-dimethylethyl)-2,2′-bipyridine-κN1,κN1′]-, chloride

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$178.00
5 G
$941.00

$178.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$178.00
5 G
$941.00

About This Item

Empirical Formula (Hill Notation):
C18H32Cl2N2NiO4
CAS Number:
Molecular Weight:
470.06
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

$178.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>300 °C

SMILES string

[Ni+2]1([NH]2=CC=C(C=C2C3=CC(=CC=[NH]31)C(C)(C)C)C(C)(C)C)(O)(O)(O)O.C

InChI key

UTKNAEDPSHMNCR-UHFFFAOYSA-J

Application

[Ni(dtbbpy)(H2O)4]Cl2 is a Ni precatalyst used in a series of Ni/photoredox dual catalysis cross-coupling reactions. These transformations include the decarboxylative coupling of oxetanyl amino acids with aryl halides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dual Photoredox/Nickel-Catalyzed Conversion of Aryl Halides to Aryl Aminooxetanes: Computational Evidence for a Substrate-Dependent Switch in Mechanism.
Kolahdouzan K, et al.
ACS Catalysis, 10(1), 405-411 (2020)
Álvaro Gutiérrez-Bonet et al.
ACS catalysis, 6(12), 8004-8008 (2016-12-19)
A Ni/photoredox dual catalytic cross-coupling is disclosed in which a diverse range of (hetero)aryl bromides are used as electrophiles, with 1,4-dihydropyridines serving as precursors to Csp3-centered alkyl radical coupling partners. The reported method is characterized by its extremely mild reaction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service