Skip to Content
Merck
All Photos(1)

Key Documents

901149

Sigma-Aldrich

2-(2-Ethoxyethoxy)ethanamine

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$490.00

$490.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$490.00

About This Item

Empirical Formula (Hill Notation):
C6H15NO2
CAS Number:
Molecular Weight:
133.19
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

$490.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

liquid

refractive index

n/D 1.4292

density

0.92 g/mL

SMILES string

NCCOCCOCC

InChI

1S/C6H15NO2/c1-2-8-5-6-9-4-3-7/h2-7H2,1H3

InChI key

KURRHYKFNUZCSJ-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F

Flash Point(C)

73.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service