Skip to Content
Merck
All Photos(2)

Key Documents

767565

Sigma-Aldrich

Poly(ethylene glycol) methyl ether amine

average Mn 500

Synonym(s):

Polyethylene glycol, Methoxypolyethylene glycol amine, PEG NH2, mPEG amine, mPEG-NH2, methoxyPEG amine

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$432.00

$432.00


Available to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 ML
$432.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$432.00


Available to ship on17 April 2025Details


Request a Bulk Order

form

liquid

Quality Level

mol wt

PEG average Mn 450 (n~10)
average Mn 500

reaction suitability

reagent type: chemical modification reagent
reactivity: carboxyl reactive

Ω-end

amine

α-end

methoxy

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

2-8°C

SMILES string

NCCOC

InChI

1S/C3H9NO/c1-5-3-2-4/h2-4H2,1H3

InChI key

ASUDFOJKTJLAIK-UHFFFAOYSA-N

Application

PEG functionalized with amine for ligation and coupling reactions.
PEG functionalized with amine for ligation and coupling reactions.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service