Skip to Content
Merck
All Photos(1)

Key Documents

768901

Sigma-Aldrich

2-[2-(2-Aminoethoxy)ethoxy]ethanol

≥96.0% (GC)

Synonym(s):

2-[2-(2-Hydroxyethoxy)ethoxy]ethylamine, 3,6,9-Trioxadecylamine, 8-Amino-3,6-dioxaoctanol, NSC 40760, T3EGMA, Triethylene glycol monoamine, Triglycolamine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$117.00
1 G
$208.00

$117.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$117.00
1 G
$208.00

About This Item

Empirical Formula (Hill Notation):
C6H15NO3
CAS Number:
Molecular Weight:
149.19
Beilstein:
1739188
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

$117.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥96.0% (GC)

form

liquid

density

1.0773 g/cm3 at 20 °C (lit.)

SMILES string

NCCOCCOCCO

InChI

1S/C6H15NO3/c7-1-3-9-5-6-10-4-2-8/h8H,1-7H2

InChI key

ASDQMECUMYIVBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service