Skip to Content
Merck
All Photos(1)

Key Documents

900865

Sigma-Aldrich

4-Formyl-2-((trimethylsilyl)ethynyl)benzenesulfonyl fluoride

≥95%

Synonym(s):

Formyl sulfonyl fluoride TMS-Alkyne, Probe building block

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13FO3SSi
CAS Number:
Molecular Weight:
284.38
UNSPSC Code:
12352200
NACRES:
NA.22
Product 900865 is not currently sold in your country. Contact Technical Service

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: click chemistry

storage temp.

−20°C

SMILES string

O=CC1=CC=C(S(=O)(F)=O)C(C#C[Si](C)(C)C)=C1

Application

4-Formyl-2-((trimethylsilyl)ethynyl)benzenesulfonyl fluoride is used for chemical probe synthesis, this trifunctional building block contains an aryl sulfonyl fluoride group, alkyne tag, and aldehyde synthetic handle. When appended to a ligand or pharmacophore through its formyl linker, this building block allows for SuFEx-enabled, context-specific covalent modification of a biological target with potential for downstream applications via the alkyne tag. Use alone or in parallel with other multi-functional building blocks to discover the optimal probe for your chemical biology experiments.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In partnership with Pfizer chemists, we have compiled a collection of trifunctional building blocks to enable development of probes.

In partnership with Pfizer chemists, we have compiled a collection of trifunctional building blocks to enable development of probes.

In collaboration with K. Barry Sharpless and coworkers, we offer a variety of sulfonyl fluoride reagents that undergo a “Click II” reaction.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service