900611
(3-Hydroxyphenyl)(4-(prop-2-yn-1-yloxy)phenyl)methanone
≥95%
Synonym(s):
Hydroxyl benzophenone alkyne, Probe building block
About This Item
Recommended Products
Assay
≥95%
form
solid
reaction suitability
reaction type: click chemistry
storage temp.
−20°C
Application
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
In partnership with Pfizer chemists, we have compiled a collection of trifunctional building blocks to enable development of probes.
In partnership with Pfizer chemists, we have compiled a collection of trifunctional building blocks to enable development of probes.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service