Skip to Content
Merck
All Photos(1)

Documents

726842

Sigma-Aldrich

(R)-(+)-4-Methoxy-α-methylbenzylamine

ChiPros®, produced by BASF, 99%

Synonym(s):

(R)-(+)-1-(4-Methoxyphenyl)ethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO
CAS Number:
Molecular Weight:
151.21
Beilstein:
2413029
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

grade

produced by BASF

Assay

≥98.5% (GC)
99%

form

liquid

optical purity

enantiomeric excess: ≥98.5%

density

1.024 g/mL at 20 °C (lit.)

SMILES string

COc1ccc(cc1)[C@@H](C)N

InChI

1S/C9H13NO/c1-7(10)8-3-5-9(11-2)6-4-8/h3-7H,10H2,1-2H3/t7-/m1/s1

InChI key

JTDGKQNNPKXKII-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

(R)-(+)-4-Methoxy-α-methylbenzylamine can be used as a reactant to prepare:
  • Enantiopure stereoisomers of hemicryptophanes, which are used for the recognition of glucopyranosides.
  • Bicyclic Geissman-Waiss lactone via intramolecular ring-closure reaction of the diastereomeric mixture of sulfonium salts.
  • N-[(1R)-1-(4-Methoxyphenyl)ethyl]-N′-methylthiourea by reacting with methyl isothiocyanate.

Legal Information

ChiPros is a registered trademark of BASF SE

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved hemicryptophane hosts for the stereoselective recognition of glucopyranosides
Schmitt A, et al.
Organic & Biomolecular Chemistry, 12(24), 4211-4217 (2014)
Synthesis of (+)-and (-)-Geissman-Waiss lactone from chiral sulfonium salts
Lopez-Gonzalez R, et al.
Tetrahedron Letters, 12(24), 151697-151697 (2020)
Yohan Park et al.
Archives of pharmacal research, 35(8), 1393-1401 (2012-09-04)
Thirty two thiourea derivatives were prepared and their agonistic activities on the retinoic acid receptor-related orphan receptor α (RORα) were evaluated. The replacement of the 3-allyl-2-imino-thiazolidin-4-one moiety of the lead compound CGP52608 (1) with various functional group substituted aromatic rings

Articles

Chiral amines play an important role in stereoselective organic synthesis. They are used directly as resolving agents, building blocks, or chiral auxiliaries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service