Merck
  • N-methylthioureas as new agonists of retinoic acid receptor-related orphan receptor.
N-methylthioureas as new agonists of retinoic acid receptor-related orphan receptor.
Archives of pharmacal research (2012-09-04)
Yohan Park, Suckchang Hong, Myungmo Lee, Hyojun Jung, Won-Jea Cho, Eun-Jin Kim, Ho-Young Son, Mi-Ock Lee, Hyeung-Geun Park
ABSTRACT
Thirty two thiourea derivatives were prepared and their agonistic activities on the retinoic acid receptor-related orphan receptor α (RORα) were evaluated. The replacement of the 3-allyl-2-imino-thiazolidin-4-one moiety of the lead compound CGP52608 (1) with various functional group substituted aromatic rings, improved the agonistic activity of RORα. Among the prepared derivatives, 1-methyl-3-(4-phenoxy-benzyl)-thiourea (32) showed 2.6-fold higher agonistic activity than CGP52608 in the RORα-activation assay.
Sigma-Aldrich
(R)-(+)-4-Methoxy-α-methylbenzylamine, ChiPros®, produced by BASF, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
CGP 52608, ≥98%, solid
Pricing and availability is not currently available.
Sigma-Aldrich
N-Methylthiourea, 97%
Pricing and availability is not currently available.