Skip to Content
Merck
All Photos(1)

Key Documents

71034

Sigma-Aldrich

Chloro-dimethyl-octadecylsilane solution

produced by Wacker Chemie AG, Burghausen, Germany, 70% in toluene

Synonym(s):

Silane ODM-Tol Sol, Dimethyl-octadecylchlorosilane, Octadecyldimethylchlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H43ClSi
CAS Number:
Molecular Weight:
347.09
Beilstein:
2246012
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

grade

produced by Wacker Chemie AG, Burghausen, Germany

form

solution

composition

n- and iso-isomer, ≥64%
n-isomer, >59%

concentration

70% in toluene

density

0.86 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC[Si](C)(C)Cl

InChI

1S/C20H43ClSi/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-22(2,3)21/h4-20H2,1-3H3

InChI key

GZGREZWGCWVAEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Choro-dimethyl-octadecylsilane can be used as a precursor for the synthesis of hydrophobic silicon nanoparticles with fluorescence properties via low temperature colloid-chemical synthetic method. It can also be used to prepare non-porous C18 silica gel stationary phase which is used in the separation of chiral enantiomers by capillary electrochromatography method.

Other Notes

prices for bulk quantities on request

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silicon nanoparticle based fluorescent biological label via low temperature thermal degradation of chloroalkylsilane
D Pradip, et al.
Nanoscale, 5(13), 5732-5737 (2013)
Preparation of 1 ?m non-porous C18 silica gel stationary phase for chiral-pressurized capillary electrochromatography
Lu Yangfang, et al.
Se Pu / Chinese Journal of Chromatography, 33(3), 209-214 (2015)
Victor K Langsi et al.
Journal of chromatography. A, 1402, 17-26 (2015-06-02)
Nanometer control over the porous shell thickness of sub-2-μm-shell particles is investigated. Three seeded growth mesoporous thin shell particles for HPLC were prepared, with 0.05μm (or 50nm) porous shell layers: particle sizes 1.5μm (solid core diameters 1.4μm), 1.7μm (solid core
Faiz Ali et al.
Journal of separation science, 37(23), 3426-3434 (2014-09-23)
Ground porous silica monolith particles with an average particle size of 2.34 μm and large pores (363 Å) exhibiting excellent chromatographic performance have been synthesized on a relatively large scale by a sophisticated sol-gel procedure. The particle size distribution was
Adelina Smirnova et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 31(11), 1201-1204 (2015-11-13)
Micro- and nanofluidics has attracted much attention, particularly concerning single-cell analysis when small amounts of liquids are examined. In present work we successfully fabricated extended-nano channels that were more narrow and shorter (2 mm) as well as wider and longer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service