Skip to Content
Merck
All Photos(1)

Key Documents

40242

Sigma-Aldrich

Chloro(dodecyl)dimethylsilane

≥95.0% (GC)

Synonym(s):

Chloro-dimethyl-dodecylsilane, Dimethyldodecylchlorosilane, Dodecyldimethylchlorosilane

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$154.00

$154.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$154.00

About This Item

Linear Formula:
H3C(CH2)11Si(CH3)2Cl
CAS Number:
Molecular Weight:
262.93
Beilstein:
5240407
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

$154.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.445

bp

291-293 °C

density

0.865 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCCCC[Si](C)(C)Cl

InChI

1S/C14H31ClSi/c1-4-5-6-7-8-9-10-11-12-13-14-16(2,3)15/h4-14H2,1-3H3

InChI key

DLLABNOCKQMTEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service