Skip to Content
Merck
All Photos(1)

Key Documents

496472

Sigma-Aldrich

Bis[4-(vinyloxymethyl)cyclohexylmethyl] glutarate

Synonym(s):

VEctomer® 4020 vinyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2[CH2CO2CH2C6H10CH2OCH=CH2]2
CAS Number:
Molecular Weight:
436.58
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

refractive index

n20/D 1.487 (lit.)

density

1.05 g/mL at 25 °C (lit.)

SMILES string

C=COCC1CCC(CC1)COC(=O)CCCC(=O)OCC2CCC(CC2)COC=C

InChI

1S/C25H40O6/c1-3-28-16-20-8-12-22(13-9-20)18-30-24(26)6-5-7-25(27)31-19-23-14-10-21(11-15-23)17-29-4-2/h3-4,20-23H,1-2,5-19H2

InChI key

SDNBHBGJJPWRJG-UHFFFAOYSA-N

Legal Information

Product of Morflex, Inc.
VEctomer is a registered trademark of Vertellus Specialties, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service