Skip to Content
Merck
All Photos(1)

Key Documents

406171

Sigma-Aldrich

1,4-Cyclohexanedimethanol divinyl ether, mixture of isomers

98%

Synonym(s):

1,4-Bis[(vinyloxy)methyl]cyclohexane

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$191.00
500 ML
$648.00

$191.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$191.00
500 ML
$648.00

About This Item

Linear Formula:
C6H10(CH2OCH=CH2)2
CAS Number:
Molecular Weight:
196.29
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$191.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.472 (lit.)

bp

126 °C/14 mmHg (lit.)

density

0.919 g/mL at 25 °C (lit.)

SMILES string

C=COCC1CCC(CC1)COC=C

InChI

1S/C12H20O2/c1-3-13-9-11-5-7-12(8-6-11)10-14-4-2/h3-4,11-12H,1-2,5-10H2

InChI key

DQNSRQYYCSXZDF-UHFFFAOYSA-N

Storage and Stability

May form peroxides

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

234.5 °F - closed cup

Flash Point(C)

112.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service