Skip to Content
Merck
All Photos(1)

Documents

409952

Sigma-Aldrich

1,4-Cyclohexanedimethanol vinyl ether, mixture of cis and trans

85%, technical grade

Synonym(s):

1,4-Bis(hydroxymethyl)cyclohexane monovinyl ether, 4-(Hydroxymethyl)cyclohexylmethyl vinyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHOCH2C6H10CH2OH
CAS Number:
Molecular Weight:
170.25
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Assay

85%

form

liquid

refractive index

n20/D 1.482 (lit.)

bp

262 °C (lit.)

mp

12 °C (lit.)

density

0.986 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OCC1CCC(CC1)COC=C

InChI

1S/C10H18O2/c1-2-12-8-10-5-3-9(7-11)4-6-10/h2,9-11H,1,3-8H2

InChI key

INRGAWUQFOBNKL-UHFFFAOYSA-N

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service