Skip to Content
Merck
All Photos(1)

Key Documents

494402

Sigma-Aldrich

Butane

99%

Synonym(s):

n-Butane

Sign Into View Organizational & Contract Pricing

Select a Size

170 G
$707.00

$707.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
170 G
$707.00

About This Item

Linear Formula:
CH3CH2CH2CH3
CAS Number:
Molecular Weight:
58.12
Beilstein:
969129
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

$707.00


Check Cart for Availability

Request a Bulk Order

vapor density

2.11 (vs air)

Quality Level

Assay

99%

form

gas

bp

−0.5 °C (lit.)

mp

−138 °C (lit.)

density

0.579 g/mL at 20 °C (lit.)

SMILES string

CCCC

InChI

1S/C4H10/c1-3-4-2/h3-4H2,1-2H3

InChI key

IJDNQMDRQITEOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butane is commonly used as a solvent for the extraction of essential oils and other compounds from plant materials.[1]

Application

Butane may be used as a test compound in combustion[2], oxidation[3] and isomerization studies.[4]

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin K Keitz et al.
Journal of the American Chemical Society, 133(40), 16277-16284 (2011-09-17)
The preparation of new phosphonium alkylidene ruthenium metathesis catalysts containing N-heterocyclic carbenes (NHCs) that result in a preference for degenerate metathesis is described. The reaction of the catalysts with ethylene or substrates relevant to ring-closing metathesis (RCM) produced ruthenacyclobutanes that
D R Williams et al.
Resuscitation, 37(1), 43-45 (1998-07-17)
We report a case of out of hospital defibrillation in a 15-year-old schoolgirl who developed ventricular fibrillation (VF) in association with butane gas inhalation. Although defibrillation performed by an attending paramedic was successful in restoring a cardiac output, her clinical
Elizabeth Barsotti et al.
Langmuir : the ACS journal of surfaces and colloids, 34(15), 4473-4483 (2018-04-04)
We use the comparison of experimentally measured isotherms for propane, n-butane, and n-pentane in 2.90, 4.19, and 8.08 nm MCM-41 to show that the current model for the progression of capillary condensation may not hold true for chain molecules, such
The role of transition metal promoters on sulfated zirconia catalysts for low-temperature butane isomerization.
Tabora JE & Davis RJ.
J. Catal., 162(1), 125-133 (1996)
Yue Deng et al.
Frontiers in microbiology, 9, 1576-1576 (2018-08-02)
To investigate the diversity of butane-oxidizing bacteria in soils contaminated by long-term light hydrocarbon microseepage and the influence of butane on the soil microbial community, a quantitative study and identification of butane-oxidizing bacteria (BOB) in soils at the Puguang gas

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service