Key Documents
Z146986
Sigma-Aldrich
PTFE sealing washers
CGA Inlet size 110
Select a Size
1 PKG
$87.70
$87.70
In StockDetails
Select a Size
Change View
1 PKG
$87.70
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
$87.70
In StockDetails
packaging
pkg of 25 ea
CGA Inlet size
110
General description
Use with Aldrich lecture-bottle regulators and control valves.
Choose from one of the most recent versions:
Certificates of Analysis (COA)