Skip to Content
Merck
All Photos(1)

Documents

481378

SAFC

Allyl chloride

Synonym(s):

3-Chloro-1-propene, Chlorallylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2Cl
CAS Number:
Molecular Weight:
76.52
Beilstein:
635704
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.6 (vs air)

Quality Level

vapor pressure

20.58 psi ( 55 °C)
5.71 psi ( 20 °C)

Assay

98.0% (returnable containers)

expl. lim.

11.2 %

refractive index

n20/D 1.414 (lit.)

bp

44-46 °C (lit.)

mp

−130 °C (lit.)

density

0.939 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClCC=C

InChI

1S/C3H5Cl/c1-2-3-4/h2H,1,3H2

InChI key

OSDWBNJEKMUWAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Nervous system,Liver,Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-25.6 °F

Flash Point(C)

-32 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shin-ichi Ikeda et al.
Chemical communications (Cambridge, England), (4)(4), 457-459 (2006-02-24)
The Ni-catalyzed coupling of allyl chlorides and enynes has been developed; the cyclization of enynes was triggered by the addition of pi-allylnickel species to the alkyne part, followed by the incorporation of the alkene part.
B M de Rooij et al.
Drug metabolism and disposition: the biological fate of chemicals, 24(7), 765-772 (1996-07-01)
Allyl chloride (AC) is used as intermediate in the synthesis of epichlorohydrin (ECH). We investigated the biotransformation of AC in rats to select potential urinary biomarkers of exposure. For this purpose, we developed analytical methods to measure different selected urinary
Justin M Chalker et al.
Chemical communications (Cambridge, England), (25)(25), 3714-3716 (2009-06-27)
Multiple, complementary methods are reported for the chemical conversion of cysteine to S-allyl cysteine on protein surfaces, a useful transformation for the exploration of olefin metathesis on proteins.
Yan-Bo Yu et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(46), 14643-14648 (2012-10-12)
Simply id(all)ylic: a copper- and phosphine-ligand-free Pd-catalyzed direct allylation of electron-deficient polyfluoroarenes with allylic chlorides and the reaction mechanism are described. The simple catalytic system, broad substrate scope, and excellent functional-group compatibility of this protocol provides a useful and facile
Carbocycle synthesis through facile and efficient palladium-catalyzed allylative de-aromatization of naphthalene and phenanthrene allyl chlorides.
Shirong Lu et al.
Angewandte Chemie (International ed. in English), 47(23), 4366-4369 (2008-04-30)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service