Skip to Content
Merck
All Photos(1)

Key Documents

470414

Sigma-Aldrich

2-Methoxyphenylmagnesium bromide solution

1.0 M in THF

Synonym(s):

(2-Methoxyphenyl)magnesium bromide, (o-Anisyl)bromomagnesium, (o-Methoxyphenyl)magnesium bromide (6CI), Bromo(2-methoxyphenyl)magnesium, Bromo(o-methoxyphenyl)magnesium, o-Anisylmagnesium bromide

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$170.00

$170.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
$170.00

About This Item

Linear Formula:
CH3OC6H4MgBr
CAS Number:
Molecular Weight:
211.34
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$170.00


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in THF

bp

65-67 °C

density

1.011 g/mL at 25 °C

SMILES string

COc1ccccc1[Mg]Br

InChI

1S/C7H7O.BrH.Mg/c1-8-7-5-3-2-4-6-7;;/h2-5H,1H3;1H;/q;;+1/p-1

InChI key

BJJFGIPCBFILPB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. How do I know the package material of 470414-100ML and 470414-1L? An ampoule, a sealed bottle, or a general pack (a bottle with a cap)?

    1 answer
    1. The 100 mL size is packaged in a 100 mL amber glass bottle. The 1 L size is packaged in a 1 L amber glass bottle. Both sizes have a Sure/Seal™ cap. To learn more about the Sure/Seal packaging system, see the link below:
      https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/organic-reaction-toolbox/sureseal

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service