Skip to Content
Merck
All Photos(1)

Key Documents

470260

Sigma-Aldrich

4-Methoxyphenylmagnesium bromide solution

0.5 M in THF

Synonym(s):

(4-Methoxyphenyl)magnesium bromide, 4-Anisylmagnesium bromide, p-Methoxyphenylmagnesium bromide, Bromo(4-methoxyphenyl)magnesium

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$147.00
1 L
$360.00

$147.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
$147.00
1 L
$360.00

About This Item

Linear Formula:
CH3OC6H4MgBr
CAS Number:
Molecular Weight:
211.34
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$147.00


Available to ship on22 April 2025Details


Request a Bulk Order

reaction suitability

reaction type: Grignard Reaction

Quality Level

concentration

0.5 M in THF

bp

65-67 °C

density

0.955 g/mL at 25 °C

SMILES string

COc1ccc([Mg]Br)cc1

InChI

1S/C7H7O.BrH.Mg/c1-8-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

JKXAZEXIOPFJSC-UHFFFAOYSA-M

Application

4-Methoxyphenylmagnesium bromide is a general Grignard reagent that can be used:
  • In the denickelation of porphyrins and to replace the metal center with magnesium.[1]
  • To synthesize a tubulin polymerization inhibitor, 2-Amino-3,4,5-trimethoxybenzophenone.[2]
  • In the total synthesis of (−)-centrolobine.[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F

Flash Point(C)

-20 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Two successive one-pot reactions leading to the expeditious synthesis of (−)-centrolobine.
Boulard L, et al.
Tetrahedron Letters, 45(35), 6603-6605 (2004)
Demetalation of Metal Porphyrins via Magnesium Porphyrins by Reaction with Grignard Reagents.
Murakami K, et al.
Chemistry?A European Journal , 19(28), 9123-9126 (2013)
2?Amino?3, 4, 5?Trimethoxybenzophenones as Potent Tubulin Polymerization Inhibitors.
Chuang H Y, et al.
ChemMedChem, 6(3), 450-456 (2011)
Ryosuke Koyama et al.
Bioorganic & medicinal chemistry, 28(11), 115492-115492 (2020-04-16)
Effective chemotherapy for solid cancers is challenging due to a limitation in permeation that prevents anticancer drugs from reaching the center of the tumor, therefore unable to limit cancer cell growth. To circumvent this issue, we planned to apply the

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service