Skip to Content
Merck
All Photos(4)

Key Documents

110299

Sigma-Aldrich

Butyl vinyl ether

contains 0.01% potassium hydroxide as stabilizer, 98%

Synonym(s):

(Butyloxy)ethylene, Vinyl butyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$77.80
500 ML
$113.00
1 L
$156.00
2 L
$347.00

$77.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$77.80
500 ML
$113.00
1 L
$156.00
2 L
$347.00

About This Item

Linear Formula:
CH3(CH2)3OCH=CH2
CAS Number:
Molecular Weight:
100.16
Beilstein:
1560217
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$77.80


Check Cart for Availability

Request a Bulk Order

vapor density

3.5 (vs air)

Quality Level

vapor pressure

42 mmHg ( 20 °C)

Assay

98%

autoignition temp.

437 °F

contains

0.01% potassium hydroxide as stabilizer

refractive index

n20/D 1.400 (lit.)

bp

94 °C (lit.)

mp

−92 °C (lit.)

density

0.774 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCOC=C

InChI

1S/C6H12O/c1-3-5-6-7-4-2/h4H,2-3,5-6H2,1H3

InChI key

UZKWTJUDCOPSNM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butyl vinyl ether (BVE) is a vinyl ether monomer that is commonly used as a reactive diluent in the radiation-curing process of hybrid polymer coatings for paints, inks, and adhesives. This is due to its high reactivity, which is attributed to the presence of its vinyl group. The reactivity of BVE allows for the polymerization and copolymerization with other monomers. It can also act as a co-monomer when combined with olefins, siloxanediols, and diisocyanates. Furthermore, it can be used to produce homopolymers suitable for applications such as adhesives, paints, lubricants, and elastomers.[1][2]

Application

Butyl vinyl ether can be used as a monomer:
  • In the synthesis of statistical copolymers through metallocene-mediated cationic polymerization. These statistical copolymers find applications in various fields which include adhesives, surface coatings, lubricants, greases, elastomers, melting compounds, fibers, and films.
  • In the synthesis of poly(vinyl ether) plastics for specific applications in the optical industry. poly(vinyl ether) plastics can be utilized in the production of high-quality optical lenses for various applications, such as eyeglasses, camera lenses, and telescopes.
  • In the synthesis of poly (butyl vinyl ether) adhesives for direct ink writing (DIW) 3D printing. The BVE-based adhesives exhibit viscoelastic properties, rheological properties, and printability which are advantageous for the additive build-up of extruded layers in DIW.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shifang Liu et al.
The Journal of organic chemistry, 71(19), 7467-7470 (2006-09-09)
meso-2,4-Bis(diphenylphosphino)pentane (mBDPP) has proved to be an effective regiocontrolling ligand for palladium-catalyzed internal arylation by aryl bromides of electron-rich olefins in a common solvent DMSO with no need for any halide scavengers. The arylation of the benchmark electron-rich olefin butyl
M Isobe et al.
The Journal of toxicological sciences, 20(2), 161-164 (1995-05-01)
4-Nitrophenyl vinyl ether (NPVE) and phenyl vinyl ether (PVE) administered i.p. in mice lowered hepatic non-protein sulfhydryl (NP-SH) content, but did not elevate the serum glutamate pyruvate transaminase (GPT) activity. n-Butyl vinyl ether (BVE) showed no significant effects either on

Questions

  1. hi is this product bottled in nitrogen or air? thanks

    1 answer
    1. The product is not packaged under nitrogen. The container should be tightly closed and the recommended storage temperature 2-8 °C.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service