Skip to Content
Merck
All Photos(1)

Key Documents

294977

Sigma-Aldrich

Air

dry

Sign Into View Organizational & Contract Pricing

Select a Size

57 L
$350.00

$350.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
57 L
$350.00

About This Item

MDL number:
UNSPSC Code:
12142100
NACRES:
NA.22

$350.00


Check Cart for Availability

Request a Bulk Order

vapor density

1 (vs air)

Quality Level

impurities

≤10 ppm water

bp

−189 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service