Skip to Content
Merck
All Photos(1)

Documents

163694

Sigma-Aldrich

2,4,6-Tribromoaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br3C6H2NH2
CAS Number:
Molecular Weight:
329.81
Beilstein:
2209258
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

bp

300 °C (lit.)

mp

120-122 °C (lit.)

solubility

alcohol: soluble (cold)(lit.)
alcohol: soluble (hot)(lit.)
chloroform: soluble(lit.)
diethyl ether: soluble(lit.)

SMILES string

Nc1c(Br)cc(Br)cc1Br

InChI

1S/C6H4Br3N/c7-3-1-4(8)6(10)5(9)2-3/h1-2H,10H2

InChI key

GVPODVKBTHCGFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Electrochemical oxidation of 2,4,6-tribromoaniline in aqueous sulphuric acid solutions at platinum electrode has been studied by rotating disc electrode and cyclic voltammetry. 2,4,6-Tribromoaniline on closed-tube pyrolysis yields polybromophenazines. It is an intermediate in the formulation of flame retardants incorporated to plastic materials.

Application

2,4,6-Tribromoaniline was used in simultaneous determination of phenolic brominated flame retardants and their by-products in water.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Voltammetric study of the oxidation kinetics of 2, 4, 6,-tribromoaniline in acid medium.
Brillas E, et al.
J. Electroanal. Chem. Interfac. Electrochem., 160(1), 185-198 (1984)
Formation of bromodioxin analogs from closed tube pyrolysis of 2, 4, 6-tribromoaniline (I).
Alsabbagh AM, et al.
Chemosphere, 17(12), 2391-2396 (1988)
P López et al.
Journal of chromatography. A, 1216(3), 334-345 (2008-09-03)
Brominated flame retardants (BFRs) are the chemicals of high importance within the REAch framework. In addition to polybrominated diphenyl ethers (PBDEs), hexabromocyclododecane (HBCD) and tetrabromobisphenol A (TBBPA), other BFRs such as bromophenols, intermediates in FR formulation like bromoanilines, and their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service