Skip to Content
Merck
All Photos(2)

Key Documents

C60007

Sigma-Aldrich

4-Chloro-3-nitrobenzoic acid

98%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$61.80

$61.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$61.80

About This Item

Linear Formula:
ClC6H3(NO2)CO2H
CAS Number:
Molecular Weight:
201.56
Beilstein:
783626
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$61.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder

mp

180-183 °C (lit.)

SMILES string

OC(=O)c1ccc(Cl)c(c1)[N+]([O-])=O

InChI

1S/C7H4ClNO4/c8-5-2-1-4(7(10)11)3-6(5)9(12)13/h1-3H,(H,10,11)

InChI key

DFXQXFGFOLXAPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elastic and bendable caffeine cocrystals: implications for the design of flexible organic materials.
Soumyajit Ghosh et al.
Angewandte Chemie (International ed. in English), 51(41), 10319-10323 (2012-09-13)
Ahmad Toufic Kabbani et al.
Chemical & pharmaceutical bulletin, 55(3), 446-450 (2007-03-03)
Copper and cobalt complexes with 4-chloro-3-nitrobenzoate (ClNBz) and the nitrogen ligands 1,3-diaminopropane (1,3-DAP) or o-phenylenediamine (o-PDA), were prepared and characterized. The complexes [Cu(ClNBz)2(1,3-DAP)] (1), [Cu(ClNBz)(o-PDA)]Cl (2), [Co(ClNBz)2(1,3-DAP)] (3) and [Co(ClNBz)2(o-PDA)2] (4) were characterized by FTIR, UV-Visible absorption, elemental analysis and
B D'eng et al.
Voprosy meditsinskoi khimii, 29(6), 113-117 (1983-11-01)
Hepatotoxic effects of 4-chloro-3-nitrobenzoic acid (x-NBA), 3-nitrobenzoic acid (NBA) and 4-chlorobenzoic acid (CBA) were studied at the doses corresponding to LD50, 1/10 LD50 and 1/50 LD50. The toxic effects were estimated by monitoring alterations in activity of protein-synthesizing system in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service