Skip to Content
Merck
All Photos(1)

Key Documents

D38405

Sigma-Aldrich

2,4-Dibromoaniline

98%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$385.00

$385.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$385.00

About This Item

Linear Formula:
Br2C6H3NH2
CAS Number:
Molecular Weight:
250.92
Beilstein:
2206653
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$385.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

crystals

mp

78-80 °C (lit.)

SMILES string

Nc1ccc(Br)cc1Br

InChI

1S/C6H5Br2N/c7-4-1-2-6(9)5(8)3-4/h1-3H,9H2

InChI key

DYSRXWYRUJCNFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4-Dibromoaniline can be used as:
  • A starting material to synthesize acetylenic amine by reacting with trimethylsilylacetylene, which is used as a ligand to prepare the bis-amido complex of Ti(IV).[1]
  • A substrate in the Pd-catalyzed ortho-selective cross-coupling reactions of dihaloarenes with Grignard reagents.[2]
  • A reactant to prepare dialkyl-substituted aminoaryl sulfides using a Grignard reagent.[3]
  • A starting material to synthesize substituted 2-mercapto benzimidazoles.[4]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Copper (I)-catalyzed synthesis of substituted 2-mercapto benzimidazoles
Murru S, et al.
The Journal of Organic Chemistry, 74(5), 2217-2220 (2009)
DHTP Ligands for the Highly Ortho-Selective, Palladium-Catalyzed Cross-Coupling of Dihaloarenes with Grignard Reagents: A Conformational Approach for Catalyst Improvement
Ishikawa S and Manabe K
Angewandte Chemie (International Edition in English), 122(4), 784-787 (2010)
A facile one-pot preparation of alkyl aminoaryl sulfides for the synthesis of GW7647 as an agonist of peroxisome proliferator-activated receptor α
Ham J, et al.
The Journal of Organic Chemistry, 71(15), 5781-5784 (2006)
Synthesis and crystal structure of (η5-C5H5)2 Ti(NH (2, 4-(C CSiMe3) 2C6H3)) 2
Zhang L, et al.
Journal of Chemical Crystallography, 35(9), 673-677 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service