Skip to Content
Merck
All Photos(1)

Key Documents

10802

Sigma-Aldrich

Apiezon® M

grease

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$814.00

$814.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$814.00

About This Item

UNSPSC Code:
12352100

$814.00


Check Cart for Availability

Request a Bulk Order

form

grease

solubility

toluene: soluble

General description

Apiezon®M constitutes the stationary phase for separation of methyl esters of fattyacids from n-pentanoic to octadecanoic acids using chromatography[1]. It is a vaccum grease[2].

Application

Apiezon®M was used in a study to characterize the photodamage to Escherichia coli in optical tweezers (traps)[2]. It was used to coat collecting substrate made up of polypropylene film in Rotating Drum Impactor for collection of size-segregated ambient aerosol samples[3].

Legal Information

Apiezon is a registered trademark of M & I Materials Ltd.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K C Neuman et al.
Biophysical journal, 77(5), 2856-2863 (1999-11-05)
Optical tweezers (infrared laser-based optical traps) have emerged as a powerful tool in molecular and cell biology. However, their usefulness has been limited, particularly in vivo, by the potential for damage to specimens resulting from the trapping laser. Relatively little
X-ray fluorescence spectrometry for high throughput analysis of atmospheric aerosol samples: The benefits of synchrotron X-rays.
Bukowiecki N, et al.
Spectrochimica Acta. Part B: Atomic Spectroscopy, 63(9), 929-938 (2008)
Gas-liquid chromatography: the separation and identification of the methyl esters of saturated and unsaturated acids from formic acid to n-octadecanoic acid.
A T JAMES et al.
The Biochemical journal, 63(1), 144-152 (1956-05-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service