Z273570
Apiezon® grease
L
Synonym(s):
Apiezon Vacuum Grease, Vacuum Grease
Select a Size
$499.00
Select a Size
About This Item
$499.00
Recommended Products
vapor pressure
7 × 10-11 mmHg ( 20 °C)
description
usage: general purpose
manufacturer/tradename
Apiezon
parameter
10-30 °C temperature (50-86 °F)
weight
50 g
Looking for similar products? Visit Product Comparison Guide
General description
Application
- low vapor pressure sealing and lubrication in a high vacuum
- manufacturing and servicing of high-performance vacuum pumps
- analytical techniques such as infrared or mass spectrometry where sample contamination must be avoided
Features and Benefits
- Free from contamination associated with silicone-based greases such as “creep” or “carry over”
- Offers excellent lubrication
- Ideal for use at ambient temperatures
- Has integrated radiation resistance
- Has powerful ‘gettering′ action, efficiently removes harmful grease and chemical impurities from metal and glass surfaces by absorption
- Soluble in hydrocarbon solvents thus making it easy to clean and remove
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service