Merck
  • A new solid phase micro extraction for simultaneous head space extraction of ultra traces of polar and non-polar compounds.
A new solid phase micro extraction for simultaneous head space extraction of ultra traces of polar and non-polar compounds.
Analytica chimica acta (2011-02-23)
Reza Alizadeh, Nahid Mashkouri Najafi, Sharmin Kharrazi
ABSTRACT
The results of the innovative study on a new stationary phase with high efficiency based on ZnO nano and micro rod coating on fused silica are reported in this paper. ZnO nanorods with a diameter in the range of 70-300 nm and the length of about 500 nm, have been grown on fused silica fibers using a hydrothermal process. The extraction properties of the fiber were investigated using headspace solid-phase microextraction (HS-SPME) mode coupled with gas chromatography-mass spectrometry detection (GC-MS) for 1,4-dichloro-nitrobenzene, biphenyl and acenaphthene. The calibration curves were linear up to 10(2)-10(7) ng L(-1) (R(2)>0.995) with detection limits of 10(-3) ng L(-1) for biphenyl and acenaphthene and 10 ng L(-1) for 1,4-dichloro-nitrobenzene. The RSD for single fiber and fiber-to-fiber were less than 7.0 and 11.5%, respectively. The high stability of the ZnO coating is proved at relatively high temperatures (up to 300°C) with a high extraction capacity and long lifespan (more than 100 times). Promising recoveries (91-102%) were obtained in environmental water samples analysis by applying the proposed technique.
Product Description
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME StableFlex fiber assortment kit, for use with manual holder
Supelco
SPME fiber assortment kit 1, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assortment kit 2, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, size 24 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 23 ga, StableFlex fiber, for use with autosampler
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 23 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 2, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 2, for use with autosampler, size 24 ga
Supelco
SPME fiber assortment kit 4, for use with manual holder, size 24 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 5, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, metal alloy fiber, for use with manual holder
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (1cm), needle size 23 ga, Autosampler, pk of 1 ×, plain gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (2cm), needle size 23 ga, Autosampler, pk of 1 ×, plain gray hub
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 23 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 23 ga, PDMS/DVB StableFlex, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber