Skip to Content
Merck
All Photos(3)

Key Documents

57334-U

Supelco

SPME Fiber Assembly

greener alternative

CAR/PDMS, for use with manual holder, Nitinol-core (NIT), 24 ga, 85 μm

Synonym(s):

Carboxen/PDMS SPME fiber assembly

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
NB.21
Pricing and availability is not currently available.

Product Name

SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber

material

StableFlex fiber
plain light blue hub

Quality Level

needle size

24 ga

packaging

pkg of 3 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

85 μm (CAR/PDMS)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

CAR/PDMS coating

application(s)

food and beverages

compatibility

(For gasses and low molecular weight compounds (MW 30 - 225).)
for use with manual holder

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

For Gases and Low Molecular Weight Compounds (MW 30-225)
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niels Bjerg Jensen et al.
Nature communications, 2, 273-273 (2011-04-21)
For more than 420 million years, plants, insects and their predators have co-evolved based on a chemical arms race including deployment of refined chemical defence systems by each player. Cyanogenic glucosides are produced by numerous plants and by some specialized
Jérémy Ratel et al.
Journal of chromatography. A, 1216(45), 7889-7898 (2009-09-29)
Animal-derived products are particularly vulnerable to contamination by volatile organic compounds (VOCs). These lipophilic substances, which are generated by an increasing number of sources, are easily transferred to the atmosphere, water, soil, and plants. They are ingested by livestock and
E Serrano et al.
Journal of chromatography. A, 1216(1), 127-133 (2008-11-29)
The objective of this paper is to investigate the potential of multiple headspace-solid-phase microextraction (MHS-SPME) for the determination of volatile compounds in complex matrix samples. A method based on MHS-SPME for the determination of around 20 volatile compounds, responsible of
Sudhir Kumar Pandey et al.
Journal of chromatography. A, 1216(28), 5436-5444 (2009-06-06)
In this study, the feasibility of GC-MS was evaluated for the quantification of odorous volatile organic compounds (VOCs) in environmental samples. These included methyl ethyl ketone, isobutyl alcohol, methyl isobutyl ketone, and butyl acetate plus benzene, toluene, and xylene (BTX).
Carlo Bicchi et al.
Journal of chromatography. A, 1218(6), 753-762 (2011-01-05)
The study compares standard addition (SA), stable isotope dilution assay (SIDA) and multiple headspace extraction (MHE) as methods to quantify furan and 2-methyl-furan in roasted coffee with HS-SPME-GC-MS, using CAR-PDMS as fibre coating, d(4)-furan as internal standard and in-fibre internal

Protocols

High performance, reliability, and reproducibility of HS-SPME in combination with GC/MS for the determination of VOCs in water was proven in an interlaboratory trial. The new ISO 17943 using HS-SPME is an improvement on existing official methods for this determination in terms of sensitivity and selectivity.

Questions

1–2 of 2 Questions  
  1. What is the pH range of the fiber in the properties section according to the 2-11 datasheet for Product 57334-U

    1 answer
    1. For optimum stability, a pH range of 2-11 is recommended.

      Helpful?

  2. Can we get pH range of fiber in properties section according to 2-11 datasheet?

    1 answer
    1. For optimum stability, a pH range of 2-11 is recommended.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service