Merck
  • A metabolite profiling approach to identify biomarkers of flavonoid intake in humans.
A metabolite profiling approach to identify biomarkers of flavonoid intake in humans.
The Journal of nutrition (2009-10-09)
Wai Mun Loke, Andrew M Jenner, Julie M Proudfoot, Allan J McKinley, Jonathan M Hodgson, Barry Halliwell, Kevin D Croft
ABSTRACT
Flavonoids are phytochemicals that are widespread in the human diet. Despite limitations in their bioavailability, experimental and epidemiological data suggest health benefits of flavonoid consumption. Valid biomarkers of flavonoid intake may be useful for estimating exposure in a range of settings. However, to date, few useful flavonoid biomarkers have been identified. In this study, we used a metabolite profiling approach to examine the aromatic and phenolic profile of plasma and urine of healthy men after oral consumption of 200 mg of the pure flavonoids, quercetin, (-)-epicatechin, and epigallocatechin gallate, which represent major flavonoid constituents in the diet. Following enzymatic hydrolysis, 71 aromatic compounds were quantified in plasma and urine at 2 and 5 h, respectively, after flavonoid ingestion. Plasma concentrations of different aromatic compounds ranged widely, from 0.01 to 10 micromol/L, with variation among volunteers. None of the aromatic compounds was significantly elevated in plasma 2 h after consumption of either flavonoid compared with water placebo. This indicates that flavonoid-derived aromatic compounds are not responsible for the acute physiological effects reported within 2 h in previous human intervention studies involving flavonoids or flavonoid-rich food consumption. These effects are more likely due to absorption of the intact flavonoid. Our urine analysis suggested that urinary 4-ethylphenol, benzoic acid, and 4-ethylbenzoic acid may be potential biomarkers of quercetin intake and 1,3,5-trimethoxybenzene, 4-O-methylgallic acid, 3-O-methylgallic acid, and gallic acid may be potential markers of epigallocatechin gallate intake. Potential biomarkers of (-)-epicatechin were not identified. These urinary biomarkers may provide an accurate indication of flavonoid exposure.
Product Description
Supelco
4-Hydroxybenzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Shikimic acid, analytical standard
Supelco
Melting point standard 121-123°C, analytical standard
Sigma-Aldrich
3-(4-Hydroxyphenyl)propionic acid, 98%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
4-Methoxyphenylacetic acid, ReagentPlus®, 99%
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
(−)-Epigallocatechin gallate, analytical standard
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
3,4-Dihydroxybenzoic acid, analytical standard
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Epigallocatechin gallate, primary reference standard
Supelco
trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Protocatechuic acid, primary reference standard
Sigma-Aldrich
3-Hydroxy-4-methoxycinnamic acid, predominantly trans, 97%
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
4-Methoxycinnamic acid, predominantly trans, 99%
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
3-Hydroxybenzoic acid, ReagentPlus®, 99%