Skip to Content
Merck
All Photos(2)

Documents

427608

Sigma-Aldrich

Benzoic acid

purified by sublimation, ≥99%

Synonym(s):

Benzenecarboxylic acid, Carboxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOH
CAS Number:
Molecular Weight:
122.12
Beilstein:
636131
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

4.21 (vs air)

Quality Level

vapor pressure

10 mmHg ( 132 °C)

Assay

≥99%

form

crystalline

autoignition temp.

1061 °F

purified by

sublimation

packaging

poly bottle of 10 g

bp

249 °C (lit.)

mp

121-125 °C (lit.)

solubility

alcohol: soluble(lit.)
benzene: soluble(lit.)
chloroform: soluble(lit.)

SMILES string

OC(=O)c1ccccc1

InChI

1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)

InChI key

WPYMKLBDIGXBTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzoic acid (BA) is an aromatic monocarboxylic acid. It is synthesized by the oxidizing toluene with atmospheric oxygen catalyzed by cobalt and manganese catalysts. It is utilized in cosmetic formulations as a pH adjuster and preservative. The elimination channels during the photodissociation of benzoic acid have been studied using multimass ion imaging techniques. Structural information and molecular constants have been obtained by microwave spectroscopy. Crystal structure study reveals that BA crystals are monoclinic with space group P21/c. Studies in bean and tomato plants have shown that BA induces stress tolerance at lower concentration.

Application

Benzoic acid (BA) may be used:
  • As a model compound in understanding the adsorption of aromatic carboxylic acids on gold surfaces.
  • As test sample in studying the photocatalytic activities of metal doped TiO2.
  • As an extracting agent of methylene blue (MB) from industrial wastewater by liquid–liquid extraction (LLE).

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extraction and recovery of methylene blue from industrial wastewater using benzoic acid as an extractant.
Muthuraman G, et al.
Journal of Hazardous Materials, 163(1), 363-369 (2009)
Photodegradation of benzoic acid over metal-doped TiO2.
Zhou J, et al.
Industrial & Engineering Chemistry Research, 45(10), 3503-3511 (2006)
Eagleson M.
Concise Encyclopedia Chemistry, 122-122 (1994)
A refinement of the benzoic acid structure at room temperature.
Bruno G and Randaccio L.
Acta Crystallographica Section B, Structural Crystallography and Crystal Chemistry, 36(7), 1711-1712 (1980)
Photodissociation dynamics of benzoic acid.
Dyakov YA, et al.
J. Chem. Phys. , 132(1), 014305-014305 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service