Skip to Content
Merck
All Photos(1)

Key Documents

03930590

Protocatechuic acid

primary reference standard

Synonym(s):

3,4-Dihydroxybenzoic acid, Protocatechuic acid

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€167.00

€167.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€167.00

About This Item

Linear Formula:
(HO)2C6H3CO2H
CAS Number:
Molecular Weight:
154.12
Beilstein:
1448841
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€167.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

mp

197-200 °C (dec.) (lit.)

application(s)

food and beverages

SMILES string

OC(=O)c1ccc(O)c(O)c1

InChI

1S/C7H6O4/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3,8-9H,(H,10,11)

InChI key

YQUVCSBJEUQKSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.
Protocatechuic acid belongs to the class of polyphenolic compounds, which can be generally isolated from the dried flower of roselle.[1][2] It can serve as a potential candidate possessing anticancer property, which can be involved in suppressing the growth of human promyelocytic leukemia HL-60 cells.[1]

Application

Reference Standard in the analysis of herbal medicinal products

Biochem/physiol Actions

Chemopreventive in several animal models of carcinogenesis. Blocks cell proliferation in the post-initiation phase.

Other Notes

This compound is commonly found in plants of the genus: allium carum cinnamomum curcuma ginkgo humulus melissa pimpinella thymus

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T H Tseng et al.
Biochemical pharmacology, 60(3), 307-315 (2000-06-17)
Hibiscus protocatechuic acid (PCA), a phenolic compound isolated from the dried flower of Hibiscus sabdariffa L. (Malvaceae), demonstrated antioxidant and antitumor promotion effects in our previous study. In the present study, Hibiscus PCA was found to inhibit the survival of
In vivo protective effect of protocatechuic acid on tert-butyl hydroperoxide-induced rat hepatotoxicity
Liu C-L, et al.
Food And Chemical Toxicology, 40, 635-641 (2002)
Saravanan Palani et al.
eLife, 10 (2021-07-17)
Actin filaments are central to numerous biological processes in all domains of life. Driven by the interplay with molecular motors, actin binding and actin modulating proteins, the actin cytoskeleton exhibits a variety of geometries. This includes structures with a curved
Kaarjel K Narayanasamy et al.
Frontiers in synaptic neuroscience, 13, 671288-671288 (2021-07-06)
The development of super-resolution microscopy (SRM) has widened our understanding of biomolecular structure and function in biological materials. Imaging multiple targets within a single area would elucidate their spatial localization relative to the cell matrix and neighboring biomolecules, revealing multi-protein
Qing-Han Gao et al.
Journal of agricultural and food chemistry, 60(38), 9642-9648 (2012-09-11)
This study investigated the changes in sugars, organic acids, α-tocopherol, β-carotene, phenolic profiles, total phenolic content (TPC), and antioxidant capacity of jujube fruits after four drying treatments (sun-, oven-, microwave- and freeze-drying). Sugar, organic acid, phenolic compounds, α-tocopherol, and β-carotene

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service