Skip to Content
Merck
All Photos(4)

Key Documents

P16621

Sigma-Aldrich

Phenylacetic acid

99%

Synonym(s):

α-Toluic acid, α-Tolylic acid, Benzeneacetic acid, PAA

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€36.40
100 G
€42.50
500 G
€68.10
1 KG
€95.90

€36.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€36.40
100 G
€42.50
500 G
€68.10
1 KG
€95.90

About This Item

Linear Formula:
C6H5CH2CO2H
CAS Number:
Molecular Weight:
136.15
Beilstein:
1099647
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€36.40


Check Cart for Availability

Request a Bulk Order

vapor density

~4 (vs air)

Quality Level

vapor pressure

1 mmHg ( 97 °C)

Assay

99%

form

crystals

bp

265 °C (lit.)

mp

76-78 °C (lit.)

density

1.081 g/mL at 25 °C (lit.)

SMILES string

OC(=O)Cc1ccccc1

InChI

1S/C8H8O2/c9-8(10)6-7-4-2-1-3-5-7/h1-5H,6H2,(H,9,10)

InChI key

WLJVXDMOQOGPHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Phenylacetic acid can be used to synthesize a variety of phenyl substituted compounds by Pd (II)-catalyzed C-H activation/aryl-aryl coupling reaction.[1][2]
  • It can be employed in the preparation of Mn16 single-molecule magnets by reductive aggregation route.[3]
  • It can be used as a precursor to synthesize chromones, isoflavones, and homoisoflavones.[4]
  • It can also be used as a key precursor in the synthesis of emethallicin E[5] and (−)-incarviatone A.[6]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and biological evaluation of epidithio-, epitetrathio-, and bis-(methylthio) diketopiperazines: synthetic methodology, enantioselective total synthesis of epicoccin G, 8, 8?-epi-ent-rostratin B, gliotoxin, gliotoxin G, emethallicin E, and haematocin and discovery of new antiviral and antimalarial agents.
Nicolaou K C, et al.
Journal of the American Chemical Society, 134(41), 17320-17332 (2012)
Enantioselective Total Synthesis of (?)-Incarviatone A.
Hong B, et al.
Journal of the American Chemical Society, 137(37), 11946-11949 (2015)
A mild and efficient protocol to synthesize chromones, isoflavones, and homoisoflavones using the complex 2, 4, 6-trichloro-1, 3, 5-triazine/dimethylformamide.
Basha G M, et al.
Canadian Journal of Chemistry, 91(8), 763-768 (2013)
A Family of Mn16 Single-Molecule Magnets from a Reductive Aggregation Route.
King P, et al.
Inorganic Chemistry, 43(23), 7315-7323 (2004)
Versatile Pd (II)-Catalyzed C? H Activation/Aryl? Aryl Coupling of Benzoic and Phenyl Acetic Acids.
Wang D H, et al.
Journal of the American Chemical Society, 130(52), 17676-17677 (2008)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service