Skip to Content
Merck
All Photos(1)

Key Documents

V900709

Sigma-Aldrich

Triphenylamine

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3N
CAS Number:
Molecular Weight:
245.32
Beilstein:
2050487
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay:
98%
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

98%

bp

347-348 °C (lit.)

mp

124-128 °C (lit.)

SMILES string

c1ccc(cc1)N(c2ccccc2)c3ccccc3

InChI

1S/C18H15N/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H

InChI key

ODHXBMXNKOYIBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiu-Qiang Huang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 154, 164-170 (2015-11-02)
A new series of trans-alkenes (3a-3e) containing different electron-donating groups were synthesized by the solvent-free Horner-Wadsworth-Emmons reaction, and characterized by infrared, hydrogen nuclear magnetic resonance, mass spectrometry and elemental analysis. Their UV-visible absorption, one-photon excited fluorescence, two-photon absorption, and two-photon
Minggang Tian et al.
Organic & biomolecular chemistry, 12(32), 6128-6133 (2014-07-06)
Cysteine (Cys) plays important roles in many physiological processes of eukaryotic cells and its detection in cells is of fundamental significance. However, glutathione (GSH), homocysteine, N-acetyl-L-cysteine and other thiols greatly hamper the detection of Cys. In particular, GSH strongly interferes
Xu Jing et al.
Journal of the American Chemical Society, 137(11), 3967-3974 (2015-03-05)
The design of artificial systems that mimic highly evolved and finely tuned natural photosynthetic systems is a subject of intensive research. We report herein a new approach to constructing supramolecular systems for the photocatalytic generation of hydrogen from water by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service