Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Linear Formula:
NH2CH2CONHCH2COOH
CAS Number:
Molecular Weight:
132.12
UNSPSC Code:
12352209
PubChem Substance ID:
EC Number:
209-127-8
Beilstein/REAXYS Number:
1765223
MDL number:
Pricing and availability is not currently available.
Product Name
Gly-Gly, Vetec™, reagent grade, 99%
grade
reagent grade
product line
Vetec™
assay
99%
form
powder
technique(s)
ligand binding assay: suitable
color
white
useful pH range
7.5-8.9
pKa (25 °C)
8.2
mp
255-260 °C
SMILES string
NCC(=O)NCC(O)=O
InChI
1S/C4H8N2O3/c5-1-3(7)6-2-4(8)9/h1-2,5H2,(H,6,7)(H,8,9)
InChI key
YMAWOPBAYDPSLA-UHFFFAOYSA-N
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under Gly-Gly
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A Chatterjee et al.
Langmuir : the ACS journal of surfaces and colloids, 28(34), 12502-12508 (2012-08-21)
The early adsorption stage of glycylglycine on Si(111)7×7 surface has been studied by scanning tunneling microscopy (STM). Filled-state imaging shows that glycylglycine adsorbs dissociatively in a bidentate fashion on two adjacent Si adatoms across a dimer wall or an adatom-restatom
Lasse Jenner et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(10), 3812-3816 (2013-02-23)
Here we present an X-ray crystallography structure of the clinically relevant tigecycline antibiotic bound to the 70S ribosome. Our structural and biochemical analysis indicate that the enhanced potency of tigecycline results from a stacking interaction with nucleobase C1054 within the
Francis Impens et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(34), 12432-12437 (2014-08-13)
SUMOylation is an essential ubiquitin-like modification involved in important biological processes in eukaryotic cells. Identification of small ubiquitin-related modifier (SUMO)-conjugated residues in proteins is critical for understanding the role of SUMOylation but remains experimentally challenging. We have set up a