Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
25 mg | Check Cart for Availability | € 1.300,00 |
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
valacyclovir
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
[n]1(c2nc(nc(c2nc1)O)N)COCCOC(=O)[C@H](N)C(C)C
InChI
1S/C13H20N6O4/c1-7(2)8(14)12(21)23-4-3-22-6-19-5-16-9-10(19)17-13(15)18-11(9)20/h5,7-8H,3-4,6,14H2,1-2H3,(H3,15,17,18,20)/t8-/m1/s1
InChI key
HDOVUKNUBWVHOX-MRVPVSSYSA-N
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename USP |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard | grade pharmaceutical primary standard |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical | application(s) pharmaceutical (small molecule) |
| format neat | format neat | format - | format neat |
| API family valacyclovir | API family valganciclovir | API family - | API family valganciclovir |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
Still not finding the right product?
Explore all of our products under D-Valacyclovir
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


