Skip to Content
Merck

Skip To

1670502

USP

Tolmetin sodium

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Tolmetin sodium salt dihydrate, 1-Methyl-5-(p-toluoyl)pyrrole-2-acetic acid sodium salt dihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mg
Check Cart for Availability
€ 407,00

About This Item

Linear Formula:
C15H14NO3Na · 2H2O
CAS Number:
Molecular Weight:
315.30
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

€ 407,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

tolmetin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].[H]O[H].[H]O[H].Cc1ccc(cc1)C(=O)c2ccc(CC([O-])=O)n2C

InChI

1S/C15H15NO3.Na.2H2O/c1-10-3-5-11(6-4-10)15(19)13-8-7-12(16(13)2)9-14(17)18;;;/h3-8H,9H2,1-2H3,(H,17,18);;2*1H2/q;+1;;/p-1

InChI key

QQILXENAYPUNEA-UHFFFAOYSA-M

Gene Information

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tolmetin sodium USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Tolmetin Sodium Capsules
  • Tolmetin Sodium Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
161463614454811269550
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

API family

tolmetin

API family

sodium salicylate

API family

monensin

API family

fenoprofen

Gene Information

human ... PTGS1(5742), PTGS2(5743)

Gene Information

-

Gene Information

-

Gene Information

-


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters