Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 20 mg | Check Cart for Availability | € 1.440,00 |
About This Item
grade
pharmaceutical primary standard
API family
timolol
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
[s]1nc(c(n1)OC(CNC(C)(C)C)COc3n[s]nc3N4CCOCC4)N2CCOCC2.OC(=O)C=CC(=O)O
InChI
1S/C19H31N7O4S2.C4H4O4/c1-19(2,3)20-12-14(30-18-16(22-32-24-18)26-6-10-28-11-7-26)13-29-17-15(21-31-23-17)25-4-8-27-9-5-25;5-3(6)1-2-4(7)8/h14,20H,4-13H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
InChI key
IZSXEVCWLMEJKJ-BTJKTKAUSA-N
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| format neat | format neat | format neat | format neat |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| API family timolol | API family timolol | API family iopamidol | API family iohexol |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. −20°C | storage temp. - |
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1667326-20MG | 04061833833612 |
