Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 50 mg | Check Cart for Availability | € 1.740,00 |
About This Item
Empirical Formula (Hill Notation):
C15H23N3O3S · 0.5C4H6O4
Molecular Weight:
384.47
NACRES:
NA.24
UNSPSC Code:
41116107
grade
pharmaceutical primary standard
API family
sumatriptan
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Sumatriptan succinate Related Compound C USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
- Sumatriptan Succinate
- Sumatriptan
- Sumatriptan Nasal Spray
- Sumatriptan Tablets
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Disclaimer
Sales restrictions may apply.
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| format neat | format neat | format neat | format neat |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| storage temp. 2-8°C | storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C |
| API family sumatriptan | API family sumatriptan | API family sumatriptan | API family sumatriptan |
Still not finding the right product?
Explore all of our products under Sumatriptan succinate Related Compound C
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 3 - Repr. 2
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sumatriptan Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(2), 4185-4185 (2020)
Sumatriptan
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 30(1), 4180-4180 (2020)
Sumatriptan Succinate
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 33(3), 4187-4187 (2020)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1642223-50MG | 04061833855362 |


