Skip to Content
Merck

Skip To

1636504

USP

Sulfathiazole

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Sulfanilamidothiazole, 4-Amino-N-(2-thiazolyl)benzenesulfonamide, N1-(2-Thiazolyl)sulfanilamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C9H9N3O2S2
CAS Number:
Molecular Weight:
255.32
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
226178
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

sulfathiazole

manufacturer/tradename

USP

mp

200-202 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Nc1ccc(cc1)S(=O)(=O)Nc2nccs2

InChI

1S/C9H9N3O2S2/c10-7-1-3-8(4-2-7)16(13,14)12-9-11-5-6-15-9/h1-6H,10H2,(H,11,12)

InChI key

JNMRHUJNCSQMMB-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulfathiazole USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S2158000469028.21112
manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

analytical standard

grade

-

format

neat

format

neat

format

neat

format

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

clinical testing

application(s)

-

API family

sulfathiazole

API family

sulfathiazole

API family

-

API family

-

mp

200-202 °C (lit.)

mp

200-202 °C (lit.)

mp

200-202 °C (lit.)

mp

199-202 °C


Still not finding the right product?

Explore all of our products under Sulfathiazole


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters