Skip to Content
Merck

Skip To

1624006

USP

Sulfacetamide sodium

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Sulfacetamide sodium salt monohydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
500 mg
Check Cart for Availability
€ 407,00

About This Item

Empirical Formula (Hill Notation):
C8H9N2NaO3S · H2O
CAS Number:
Molecular Weight:
254.24
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

€ 407,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

sulfacetamide

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

NC1=CC=C(S(N([Na])C(C)=O)(=O)=O)C=C1.O

InChI

1S/C8H10N2O3S.Na.H2O/c1-6(11)10-14(12,13)8-4-2-7(9)3-5-8;;/h2-5H,9H2,1H3,(H,10,11);;1H2/q;+1;/p-1

InChI key

IHCDKJZZFOUARO-UHFFFAOYSA-M

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulfacetamide sodium USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Sulfacetamide Sodium Ophthalmic Ointment
  • Sulfacetamide Sodium Ophthalmic Solution
  • Sulfacetamide Sodium Topical Suspension
  • Sulfacetamide Sodium and Prednisolone Acetate Ophthalmic Ointment
  • Sulfacetamide Sodium and Prednisolone Acetate Ophthalmic Suspension

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1519S17000001189009
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

EDQM

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

API family

sulfacetamide

API family

sulfacetamide

API family

sulfacetamide

API family

dicloxacillin


Still not finding the right product?

Explore all of our products under Sulfacetamide sodium


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1624006-500MG04061838747013

Questions

Reviews

No rating value

Active Filters