Skip to Content
Merck

Skip To

1605056

USP

RoflumilastRelated Compound D

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3-(Cyclopropylmethoxy)-4-(difluoromethoxy)benzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
50 mg
Check Cart for Availability
€ 1.260,00

About This Item

Empirical Formula (Hill Notation):
C12H12F2O4
CAS Number:
Molecular Weight:
258.22
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:

€ 1.260,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

roflumilast

packaging

pkg of 50 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

FC(F)OC1=CC=C(C=C1OCC2CC2)C(O)=O

InChI

1S/C12H12F2O4/c13-12(14)18-9-4-3-8(11(15)16)5-10(9)17-6-7-1-2-7/h3-5,7,12H,1-2,6H2,(H,15,16)

InChI key

IGFDIFLMMLWKKY-UHFFFAOYSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
160502316050341605045
application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

packaging

pkg of 50 mg

packaging

pkg of 30 mg

packaging

pkg of 30 mg

packaging

pkg of 50 mg

API family

roflumilast

API family

roflumilast

API family

roflumilast

API family

roflumilast


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1605056-50MG04065266171587

Questions

Reviews

No rating value

Active Filters